Merck
Novel styrylbenzene derivatives for detecting amyloid deposits.
Clinica chimica acta; international journal of clinical chemistry (2014-05-13)
Manabu Nakazono, Konen Obayashi, Kazumi Sasamoto, Katsumi Tomiyoshi, Genki Suenaga, Yukio Ando
ABSTRACT
Various styrylbenzene compounds were synthesized and evaluated as mainly Aβ amyloid sensors. These compounds, however, cannot be used for detecting amyloid deposition in peripheral nerves because of the inherent sensitivity of the compounds. These compounds often generate false positives especially in the basement membrane of blood vessels in histochemical studies. To overcome these problems, we must first synthesize other styryl compounds for detecting amyloid fibrils in tissues. A wide variety of symmetrical and unsymmetrical styrylbenzene derivatives were synthesized and then these compounds were used to detect amyloid fibrils in autopsy and biopsy samples from patients with various systemic and localized forms of amyloidosis such as familial amyloidotic polyneuropathy (FAP), senile systemic amyloidosis (SSA), amyloid A (AA) amyloidosis, localized AL amyloidosis, and Alzheimer's disease. 1-Methoxy-2,5-bis-styrylbenzene and 2-(2-(2-fluoroethoxy)ethoxy)ethoxy)-2,5-bis-styrylbenzene (EEEFSB) detected amyloid fibrils in both in vitro and in vivo histopathological studies. 1-Methoxy-2,5-bis-styrylbenzene also showed a high strength of fluorescence with amyloid deposition in peripheral nerves in a patient with FAP. 1-Methoxy-2,5-bis-styrylbenzene and EEEFSB may prove a useful tool for diagnosing amyloidosis, not only in a histochemical study but also in whole body amyloid positron emission tomography (PET) imaging.
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
2-Naphthylboronic acid, ≥95.0%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Luperox® A98, Benzoyl peroxide, reagent grade, ≥98%
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Supelco
Carbon tetrachloride, analytical standard
Sigma-Aldrich
VenPure® SF, powder
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbon tetrachloride, reagent grade, 99.9%
Supelco
Carbon tetrachloride, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tin(IV) chloride, 99.995% trace metals basis
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Tin(IV) chloride, 98%
Sigma-Aldrich
3-Formylbenzoic acid, 97%