Merck
  • Pharmacokinetic study of 14-(3-methylbenzyl)matrine and 14-(4-methylbenzyl)matrine in rat plasma using liquid chromatography-tandem mass spectrometry.
Pharmacokinetic study of 14-(3-methylbenzyl)matrine and 14-(4-methylbenzyl)matrine in rat plasma using liquid chromatography-tandem mass spectrometry.
PloS one (2015-02-26)
Minjie Jiang, Lisheng Wang, Shulin Huang, Liba Xu, Chao Hu, Weizhe Jiang
ABSTRACT
A rapid, sensitive and selective high-performance liquid chromatography-tandem mass spectrometric method (HPLC-MS) was developed and validated to determine the 14-(3-methylbenzyl)matrine (3MBM) and 14-(4-methylbenzyl)matrine (4MBM) levels in rat plasma in the present study. The analytes were separated using a C18 column (1.9 μm, 2.1 mm × 100 mm) equipped with a Security Guard C18 column (5 μm, 2.1 mm × 10 mm), followed by detection via triple-quadrupole mass spectrometry using an electrospray ionization (ESI) source. Sample pretreatment involved one-step protein precipitation with isopropanol:ethyl acetate (v/v, 25:75), and pseudoephedrine hydrochloride was used as an internal standard. The method was linear in the concentration range of 5-2000 ng/ml for both compounds. The intra-day and inter-day relative standard deviations (RSDs) were less than 15%, and all relative errors (REs) were within 15%. The proposed method enables the unambiguous identification and quantification of these two compounds in vivo. This study is the first to determine the 3MBM and 4MBM levels in rat plasma after oral administration of these compounds. These results provide a meaningful basis for evaluating the clinical applications of these medicines.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
Acetonitrile, analytical standard
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%