Merck
  • Chemical and bioanalytical assessments on drinking water treatments by quaternized magnetic microspheres.
Chemical and bioanalytical assessments on drinking water treatments by quaternized magnetic microspheres.
Journal of hazardous materials (2014-12-08)
Peng Shi, Rong Ma, Qing Zhou, Aimin Li, Bing Wu, Yu Miao, Xun Chen, Xuxiang Zhang
ABSTRACT
This study aimed to compare the toxicity reduction performance of conventional drinking water treatment (CT) and a treatment (NT) with quaternized magnetic microspheres (NDMP) based on chemical analyses. Fluorescence excitation-emission-matrix combined with parallel factor analysis identified four components in source water of different rivers or lake, and the abundance of each component differed greatly among the different samples. Compared with the CT, the NT evidently reduced the concentrations of dissolved organic carbon, adsorbable organic halogens (AOX), bromide and disinfection by-products. Toxicological evaluation indicated that the NT completely eliminated the cytotoxicity, and greatly reduced the genotoxicity and oxidative stress of all raw water. In contrast, the CT increased the cytotoxicity of Taihu Lake and the Zhongshan River water, genotoxicity of Taihu Lake and the Mangshe River water, as well as the levels of superoxide dismutase and malondialdehyde of the Mangshe River water. Correlation analysis indicated that the AOX of the treated samples was significantly correlated with the genotoxicity and glutathione concentration, but exhibited no correlation with either of them for all the samples. As it can effectively reduce pollutant levels and the toxicities of drinking water, NDMP might be widely used for drinking water treatment in future.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Supelco
Dichloroacetic acid, PESTANAL®, analytical standard
Supelco
Chloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Bromoacetic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Bromoacetic acid, reagent grade, 97%
Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroacetic acid, 99%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%