Merck
  • Metabolomic serum profiling detects early-stage high-grade serous ovarian cancer in a mouse model.
Metabolomic serum profiling detects early-stage high-grade serous ovarian cancer in a mouse model.
Journal of proteome research (2015-01-09)
Christina M Jones, María Eugenia Monge, Jaeyeon Kim, Martin M Matzuk, Facundo M Fernández
ABSTRACT
Ovarian cancer is a deadly disease killing more than any other gynecologic cancer. Nonspecific symptoms, combined with a lack of early detection methods, contribute to late diagnosis and low five-year survival rates. High-grade serous carcinoma (HGSC) is the most common and deadliest subtype that results in 90% of ovarian cancer deaths. To investigate metabolic patterns for early detection of this deadly ovarian cancer, Dicer-Pten double knockout (DKO) mice that phenocopy many of the features of metastatic HGSC observed in women were studied. Using ultraperformance liquid chromatography-mass spectrometry (UPLC-MS), serum samples from 14 early-stage tumor (ET) DKO mice and 11 controls were analyzed in depth to screen for metabolic signatures capable of differentiating early-stage HGSC from controls. Iterative multivariate classification selected 18 metabolites that, when considered as a panel, yielded 100% accuracy, sensitivity, and specificity for classification. Altered metabolic pathways reflected in that panel included those of fatty acids, bile acids, glycerophospholipids, peptides, and some dietary phytochemicals. These alterations revealed impacts to cellular energy storage and membrane stability, as well as changes in defenses against oxidative stress, shedding new light on the metabolic alterations associated with early ovarian cancer stages.
Sigma-Aldrich
Suberic acid, 98%
Sigma-Aldrich
Suberic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
2-Deoxy-D-galactose, 98%
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Ricinoleic acid, analytical standard
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
Arg-Gly-Asp, ≥97% (TLC)
Sigma-Aldrich
Ricinoleic acid, ≥95%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%