Merck
  • Antagonism by salvianolic acid B of lipopolysaccharide-induced disseminated intravascular coagulation in rabbits.
Antagonism by salvianolic acid B of lipopolysaccharide-induced disseminated intravascular coagulation in rabbits.
Clinical and experimental pharmacology & physiology (2014-04-18)
Zheng Wu, Jian-Nan Li, Zhi-Quan Bai, Xi Lin
ABSTRACT
The aim of the present study was to investigate the effects of salvianolic acid B on lipopolysaccharide (LPS)-induced disseminated intravascular coagulation (DIC) in rabbits. Continuous infusion of LPS was used to induce a DIC model in rabbits. Treatment with salvianolic acid B (1, 3 or 6 mg/kg) was started simultaneously with LPS infusion (0.5 mg/kg LPS in 60 mL saline; 10 mL/h over a period of 6 h) through the contralateral marginal ear vein. Activated partial thromboplastin time (APTT), prothrombin time (PT), platelet count and fibrinogen concentration were determined, as were plasma levels of fibrin-fibrinogen degradation products (FDP), alanine aminotransferase (ALT), blood urea nitrogen (BUN), protein C activity, antithrombin III (ATIII) and tumour necrosis factor (TNF)-α concentration. The gradual impairment of haemostatic parameters was induced by continuous infusion of LPS. There were marked increases in APTT, PT, BUN, ALT and plasma TNF-α and marked decreases in the platelet count, fibrinogen, FDP, protein C and ATIII. The intravenous administration of 1, 3 or 6 mg/kg salvianolic acid B attenuated the increases in APTT, PT, BUN, ALT and plasma TNF-α and the decreases in fibrinogen, platelet, FDP, protein C and ATIII induced by LPS infusion. These observations indicate that salvianolic acid B has an effect against LPS-induced DIC in rabbits.
Sigma-Aldrich
Salvianolic acid B, ≥94% (HPLC)
Supelco
Salvianolic acid B, analytical standard
Salvianolic acid B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%