Merck
  • The effect of hyperpolarization-activated cyclic nucleotide-gated ion channel inhibitors on the vagal control of guinea pig airway smooth muscle tone.
The effect of hyperpolarization-activated cyclic nucleotide-gated ion channel inhibitors on the vagal control of guinea pig airway smooth muscle tone.
British journal of pharmacology (2014-04-26)
Alice E McGovern, Jed Robusto, Joanna Rakoczy, David G Simmons, Simon Phipps, Stuart B Mazzone
ABSTRACT
Subtypes of the hyperpolarization-activated cyclic nucleotide-gated (HCN) family of cation channels are widely expressed on nerves and smooth muscle cells in many organ systems, where they serve to regulate membrane excitability. Here we have assessed whether HCN channel inhibitors alter the function of airway smooth muscle or the neurons that regulate airway smooth muscle tone. The effects of the HCN channel inhibitors ZD7288, zatebradine and Cs(+) were assessed on agonist and nerve stimulation-evoked changes in guinea pig airway smooth muscle tone using tracheal strips in vitro, an innervated tracheal tube preparation ex vivo or in anaesthetized mechanically ventilated guinea pigs in vivo. HCN channel expression in airway nerves was assessed using immunohistochemistry, PCR and in situ hybridization. HCN channel inhibition did not alter airway smooth muscle reactivity in vitro to exogenously administered smooth muscle spasmogens, but significantly potentiated smooth muscle contraction evoked by the sensory nerve stimulant capsaicin and electrical field stimulation of parasympathetic cholinergic postganglionic neurons. Sensory nerve hyperresponsiveness was also evident in in vivo following HCN channel blockade. Cs(+) , but not ZD7288, potentiated preganglionic nerve-dependent airway contractions and over time induced autorhythmic preganglionic nerve activity, which was not mimicked by inhibitors of potassium channels. HCN channel expression was most evident in vagal sensory ganglia and airway nerve fibres. HCN channel inhibitors had a previously unrecognized effect on the neural regulation of airway smooth muscle tone, which may have implications for some patients receiving HCN channel inhibitors for therapeutic purposes.
Sigma-Aldrich
Ethanol, anhydrous, denatured
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Capsaicin, natural
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Succinylcholine chloride dihydrate, 98.0-102.0%, solid
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Capsaicin, analytical standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Trolamine, European Pharmacopoeia (EP) Reference Standard
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Supelco
Histamine, analytical standard
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Urethane, ≥99.0% (GC)