Merck
Selenitetriglycerides-Redox-active agents.
Pharmacological reports : PR (2015-01-07)
Anna Flis, Piotr Suchocki, Monika Anna Królikowska, Zofia Suchocka, Małgorzata Remiszewska, Lidia Śliwka, Iza Książek, Karolina Sitarz, Małgorzata Sochacka, Grażyna Hoser, Elżbieta Anuszewska, Piotr Wroczyński, Zenon Jastrzębski
ABSTRACT
Human prostate cancer (hPCa) is the most commonly diagnosed cancer in elderly men and is the second leading cause of male cancer death. Data from epidemiological, eco-environmental, nutritional prevention and clinical trials suggest that selenium Se(IV) can prevent prostate cancer. Selol, a new organic semisynthetic derivative of Se(IV), is a mixture of selenitetriglycerides. This mixture is non-toxic and non-mutagenic, and after po treatment - 56-times less toxic (in mice) than sodium selenite. It exhibits strong anti-cancer activity in vitro in many cancer cell lines and can overcome the cell resistance to doxorubicin. Selol seems a promising compound for prostate cancer therapy. The aim of the present study is the evaluation of Selol's influence on intracellular redox state (Eh) of prostatic tumors and the liver in androgen-dependent hPCa-bearing mice, and extracellular redox state in serum of these mice. The anticancer activity of Selol involves perturbation of the redox regulation in the androgen dependent hPCa (LNCaP) cells, but not in healthy cells. After Selol treatment, intracellular Eh has increased in tumors from -223 mV to -175 mV, while in serum it has decreased (-82 mV vs -113 mV). It shows significant changes Eh in the extra- and intracellular environment. The difference decreases from 141 mV to 62 mV. The changes suggest that a tumor cell was probably directed toward apoptosis. This is exemplified in a significant decrease in cancer tumor mass by approx. 17% after the three weeks of Selol administration.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioXtra, ≥98.0% (T)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O