Merck
  • Development of a targeted method for twenty-three metabolites related to polyphenol gut microbial metabolism in biological samples, using SPE and UHPLC-ESI-MS/MS.
Development of a targeted method for twenty-three metabolites related to polyphenol gut microbial metabolism in biological samples, using SPE and UHPLC-ESI-MS/MS.
Talanta (2014-07-26)
Mattia Gasperotti, Domenico Masuero, Graziano Guella, Fulvio Mattivi, Urska Vrhovsek
ABSTRACT
An increasing number of studies have concerned the profiling of polyphenol microbial metabolites, especially in urine or plasma, but only a few have regarded their accurate quantification. This study reports on a new ultra-performance liquid chromatography tandem mass spectrometry method with electrospray ionisation (UHPLC-ESI-MS/MS) using a simple clean-up step with solid phase extraction (SPE) and validation on different biological matrices. The method was tested with spiked samples of liver, heart, kidneys, brain, blood and urine. The purification procedure, after the evaluation of three different cartridges, makes it possible to obtain cleaner samples and better quantification of putative trace metabolites, especially related to dietary studies, with concentrations below ng/g in tissue and for urine and blood, starting from ng/ml. Limits of detection and linear range were also assessed using mixed polyphenol metabolite standards. Short chromatographic separation was carried out for 23 target compounds related to the polyphenol microbial metabolism, coupled with a triple quadrupole mass spectrometer for their accurate quantification. By analysing different spiked biological samples we were able to test metabolite detection in the matrix and validate the overall recovery of the method, from purification to quantification. The method developed can be successfully applied and is suitable for high-throughput targeted metabolomics analysis related to nutritional intervention, or the study of the metabolic mechanism in response to a polyphenol-rich diet.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
trans-3-Hydroxycinnamic acid, 99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Homovanillic acid, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)