Merck
Axial thermal gradients in microchip gas chromatography.
Journal of chromatography. A (2014-12-06)
Anzi Wang, Sampo Hynynen, Aaron R Hawkins, Samuel E Tolley, H Dennis Tolley, Milton L Lee
ABSTRACT
Fabrication technologies for microelectromechanical systems (MEMS) allow miniaturization of conventional benchtop gas chromatography (GC) to portable, palm-sized microfabricated GC (μGC) devices, which are suitable for on-site chemical analysis and remote sensing. The separation performance of μGC systems, however, has not been on par with conventional GC. Column efficiency, peak symmetry and resolution are often compromised by column defects and non-ideal injections. The relatively low performance of μGC devices has impeded their further commercialization and broader application. In this work, the separation performance of μGC columns was improved by incorporating thermal gradient gas chromatography (TGGC). The analysis time was ∼20% shorter for TGGC separations compared to conventional temperature-programmed GC (TPGC) when a wide sample band was introduced into the column. Up to 50% reduction in peak tailing was observed for polar analytes, which improved their resolution. The signal-to-noise ratios (S/N) of late-eluting peaks were increased by 3-4 fold. The unique focusing effect of TGGC overcomes many of the previous shortcomings inherent in μGC analyses.
Product Description
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
Sigma-Aldrich
(±)-2-Octanol, ≥96.0% (GC)
Sigma-Aldrich
(±)-2-Octanol, ReagentPlus®, ≥99.5% (GC)
Supelco
Pentadecane, analytical standard
Supelco
Dodecane, analytical standard
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)
Supelco
Pentane, analytical standard
Sigma-Aldrich
Tributyl phosphate, ≥99%
Sigma-Aldrich
Pentadecane, ≥99%
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
Carbon disulfide, anhydrous, ≥99%
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%
Sigma-Aldrich
Tributyl phosphate, 97%
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Sigma-Aldrich
Pentane, spectrophotometric grade, ≥99%
Sigma-Aldrich
Pentane, ≥99% (GC)
Sigma-Aldrich
Dodecane, ReagentPlus®, ≥99%
Sigma-Aldrich
Pentane, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Carbon disulfide, ReagentPlus®, low benzene, ≥99.9%
Sigma-Aldrich
Carbon disulfide, ReagentPlus®, purified by redistillation, ≥99.9%
Sigma-Aldrich
Pentane, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Carbon disulfide, suitable for HPLC, ≥99.9%
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-2-Octanol, analytical standard
Sigma-Aldrich
Pentane, reagent grade, 98%
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Supelco
Decane, analytical standard