Skip to Content
Merck
All Photos(1)

Key Documents

34956

Sigma-Aldrich

Pentane

suitable for HPLC, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.06

vapor density

2.48 (vs air)

Quality Level

vapor pressure

26.98 psi

Assay

≥99.0%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

technique(s)

HPLC: suitable

impurities

≤0.0005% free acid (as CH3COOH)
≤0.0005% non-volatile matter
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm
<0.005% water (Karl Fischer)

evapn. residue

<0.0005%

transmittance

200 nm, ≥20%
210 nm, ≥60%
220 nm, ≥85%
230 nm, ≥95%
250 nm, ≥99%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

λ

neat

UV absorption

λ: 200 nm Amax: ≤0.70
λ: 210 nm Amax: ≤0.20
λ: 220 nm Amax: ≤0.07
λ: 230 nm Amax: ≤0.02
λ: 250 nm Amax: ≤0.004

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane (n-pentane) is a hydrocarbon. Its hydroisomerization using a nanocrystalline beta (BEA) zeolite has been proposed. It has been reported as one of the products formed from the peroxidation of unsaturated fatty acid.

Application


  • Catalytic Cracking to Light Olefins: Pentane is extensively used in studies focused on catalytic cracking processes. Recent research has explored the use of extra-framework aluminum in ZSM-5 zeolites to enhance the production of light olefins from pentane. This application is crucial for improving the efficiency of catalytic processes and the production of valuable petrochemical feedstocks (He et al., Journal of Analytical and Applied Pyrolysis, 2022).

Other Notes

Important notice
The article number 34956-4X2.5L will be discontinued. Please order the single bottle 34956-2.5L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Svensson et al.
Carbohydrate polymers, 92(1), 775-783 (2012-12-12)
Dry ultra-porous cellulose fibres were obtained using a liquid exchange procedure in which water was replaced in the following order: water, methanol, acetone, and finally pentane; thereafter, the fibres were dried with Ar(g). The dry samples (of TEMPO-oxidized dissolving pulp)
Keisuke Maruyama et al.
Bioorganic & medicinal chemistry letters, 20(22), 6661-6666 (2010-10-05)
We have proposed a multi-template approach for drug development, focusing on similar fold structures of proteins, and have effectively generated lead compounds for several drug targets. Modification of these polypharmacological lead compounds is then needed to generate target-selective compounds. In
Assessment of lipid peroxidation in newborn infants and rabbits by measurements of expired ethane and pentane: influence of parenteral lipid infusion.
Wispe JR, et al.
Pediatric Research, 19(4), 374-379 (1985)
James Clarke et al.
Nature nanotechnology, 4(4), 265-270 (2009-04-08)
A single-molecule method for sequencing DNA that does not require fluorescent labelling could reduce costs and increase sequencing speeds. An exonuclease enzyme might be used to cleave individual nucleotide molecules from the DNA, and when coupled to an appropriate detection
Fuming Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 27(15), 9174-9181 (2011-07-01)
Graphene oxide (GO) can be viewed as an amphiphilic soft material, which form thin films at organic solvent-water interfaces. However, organic solvent evaporation provides little driving force, which results in slow GO transfer in aqueous phase, thus dawdling GO film

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service