Merck
  • Down-regulation of eIF4GII by miR-520c-3p represses diffuse large B cell lymphoma development.
Down-regulation of eIF4GII by miR-520c-3p represses diffuse large B cell lymphoma development.
PLoS genetics (2014-02-06)
Krystyna Mazan-Mamczarz, X Frank Zhao, Bojie Dai, James J Steinhardt, Raymond J Peroutka, Kimberly L Berk, Ari L Landon, Mariola Sadowska, Yongqing Zhang, Elin Lehrmann, Kevin G Becker, Rita Shaknovich, Zhenqiu Liu, Ronald B Gartenhaus
ABSTRACT
Deregulation of the translational machinery is emerging as a critical contributor to cancer development. The contribution of microRNAs in translational gene control has been established however; the role of microRNAs in disrupting the cap-dependent translation regulation complex has not been previously described. Here, we established that elevated miR-520c-3p represses global translation, cell proliferation and initiates premature senescence in HeLa and DLBCL cells. Moreover, we demonstrate that miR-520c-3p directly targets translation initiation factor, eIF4GII mRNA and negatively regulates eIF4GII protein synthesis. miR-520c-3p overexpression diminishes cells colony formation and reduces tumor growth in a human xenograft mouse model. Consequently, downregulation of eIF4GII by siRNA decreases translation, cell proliferation and ability to form colonies, as well as induces cellular senescence. In vitro and in vivo findings were further validated in patient samples; DLBCL primary cells demonstrated low miR-520c-3p levels with reciprocally up-regulated eIF4GII protein expression. Our results provide evidence that the tumor suppressor effect of miR-520c-3p is mediated through repression of translation while inducing senescence and that eIF4GII is a key effector of this anti-tumor activity.
Sigma-Aldrich
Anti-EIF4G3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-EIF4G3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Monoclonal Anti-EIF4G3 antibody produced in mouse, clone 1H5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-EIF4G3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
Sigma-Aldrich
MISSION® esiRNA, targeting human EIF4G3
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
ANTI-EIF4G3 antibody produced in mouse, clone 1D7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Eif4g3
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture