Merck
  • Simultaneous determination of 14-thienyl methylene matrine and matrine in rat plasma by high-performance liquid chromatography-tandem mass spectrometry and its application in a pharmacokinetic study.
Simultaneous determination of 14-thienyl methylene matrine and matrine in rat plasma by high-performance liquid chromatography-tandem mass spectrometry and its application in a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Minjie Jiang, Lisheng Wang, Weizhe Jiang, Shulin Huang
ABSTRACT
A rapid, sensitive and selective high-performance liquid chromatography-tandem mass spectrometric method (HPLC-MS) has been developed and validated for the simultaneous determination of 14-thienyl methylene matrine (TMM) and matrine (MT) in rat plasma in the present study. The analytes were separated on a C18 column (1.9 μm, 2.1 mm × 100 mm) with a security guard C18 column (5 μm, 2.1 mm × 10 mm) and a triple-quadrupole mass spectrometry equipped with an electrospray ionization (ESI) source was applied for detection. With pseudoephedrine hydrochloride as internal standard, sample pretreatment involved in a one-step protein precipitation with isopropanol:ethyl acetate (v/v, 20:80). The method was linear over the concentration ranges of 5-1000 ng/ml for TMM and 10-2000 ng/ml for MT. The intra-day and inter-day relative standard deviations (RSD) were less than 15% and the relative errors (RE) were all within 15%. The proposed method enables unambiguous identification and quantification of TMM and MT in vivo. This was the first report on determination of the TMM and MT in rat plasma after oral administration of TMM. The results provided a meaningful basis for evaluating the clinical applications of the medicine.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
Acetonitrile, analytical standard
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%