Merck
  • Breaking aggregation in a tetrathiafulvalene-fused zinc porphyrin by metal-ligand coordination to form a donor-acceptor hybrid for ultrafast charge separation and charge stabilization.
Breaking aggregation in a tetrathiafulvalene-fused zinc porphyrin by metal-ligand coordination to form a donor-acceptor hybrid for ultrafast charge separation and charge stabilization.
Dalton transactions (Cambridge, England : 2003) (2014-11-11)
Atanu Jana, Habtom B Gobeze, Masatoshi Ishida, Toshiyuki Mori, Katsuhiko Ariga, Jonathan P Hill, Francis D'Souza
ABSTRACT
A novel electron rich, tetrathiafulvalene fused zinc porphyrin, (TTF)4PZn, has been newly synthesized and characterized using spectral and electrochemical methods. In spite of the presence of eight t-butyl groups, (TTF)4PZn exhibited appreciable aggregation in solution. Scanning electron microscopic (SEM) imaging of the aggregates revealed their spherical particulate morphology. Attenuation of intermolecular aggregation was possible by metal-ligand coordination of a nitrogenous ligand. Further, using this strategy, a donor-acceptor hybrid was formed by coordinating imidazole functionalized fullerene as an electron acceptor. The occurrence of intrasupramolecular ultrafast photoinduced charge separation has been established using fluorescence and transient absorption spectroscopic techniques. The determined rate of charge separation, kCS, and rate of charge recombination, kCR were found to be 1.4 × 10(11) s(-1) and 2.5 × 10(6) s(-1), respectively. The lower kCR values indicate charge stabilization in the assembled donor-acceptor conjugate via an electron transfer-hole transfer mechanism.
Product Description
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Supelco
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, ≥98%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, ≥99.5%