Merck
  • Mechanism of fibroblast inflammatory responses to Pseudomonas aeruginosa elastase.
Mechanism of fibroblast inflammatory responses to Pseudomonas aeruginosa elastase.
Microbiology (Reading, England) (2014-01-05)
Ali O Azghani, Kourtney Neal, Steven Idell, Rodolfo Amaro, Jason W Baker, Abdelwahab Omri, Usha R Pendurthi
ABSTRACT
Receptor tyrosine kinases, including the epidermal growth factor receptors (EGFR), are able to activate the mitogen-activated protein kinases (MAPK) via several adaptor proteins and protein kinases such as Raf. EGFR can be activated by a variety of extracellular stimuli including neutrophil elastase, but we are aware of no report as to whether Pseudomonas aeruginosa produced elastase (PE) could elicit such signalling through EGFR activation. We sought to test the inference that PE modulates inflammatory responses in human lung fibroblasts and that the process occurs by activation of the EGFR/MAPK pathways. We utilized IL-8 cytokine expression as a pathway-specific end point measure of the fibroblast inflammatory response to PE. Western blot analysis was performed to detect phosphorylation of EGFR and signal transduction intermediates. Northern blot, real-time PCR, and ELISA methods were utilized to determine cytokine gene expression levels. We found that PE induces phosphorylation of the EGFR and the extracellular signal-regulated proteins (ERK1/2) of the MAPK pathway, and nuclear translocation of NF-κB. Furthermore, enzymically active PE enhances IL-8 mRNA and protein secretion. Pretreatment of the cells with specific inhibitors of EGFR, MAPK kinase and NF-κB markedly attenuated the PE-induced signal proteins phosphorylation and IL-8 gene expression and protein secretion. Collectively, the data show that PE produced by Pseudomonas aeruginosa can modulate lung inflammation by exploiting the EGFR/ERK signalling cascades and enhancing IL-8 production in the lungs via NF-κB activation.
Product Description
Sorbitol, European Pharmacopoeia (EP) Reference Standard
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Mouse Osteopontin ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human Osteopontin ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, FCC, FG