Skip to Content
Merck
All Photos(1)

Key Documents

309532

Sigma-Aldrich

Sorbitol F solution

70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides

Synonym(s):

D-Sorbitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

form

viscous liquid

concentration

70 wt. % in H2O

color

colorless

refractive index

n20/D 1.46

density

1.28 g/mL at 25 °C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • The Improved Properties of Carboxymethyl Bacterial Cellulose Films with Thickening and Plasticizing.: Research demonstrates the effectiveness of sorbitol F solution in improving the mechanical and barrier properties of carboxymethyl bacterial cellulose films. This is crucial for developing advanced biodegradable packaging materials that meet environmental and performance criteria (Sun Z et al., 2022).

  • Elaboration of hemicellulose-based films: Impact of the extraction process from spruce wood on the film properties.: Investigates the use of sorbitol F solution in the production of hemicellulose-based films, emphasizing its effectiveness in influencing film properties such as flexibility, water resistance, and biodegradability, key for sustainable packaging solutions (Chadni M et al., 2020).

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Molino et al.
The Journal of laboratory and clinical medicine, 131(5), 393-405 (1998-05-30)
D-Sorbitol (SOR) is safe, is easy to measure, and has an exceptionally high extraction ratio in the normal liver of 0.93+/-0.05 (mean+/-SD). Together with the general interest in hepatic hemodynamics, these facts motivated us to review the usefulness of this
Reddy Ranjith K Sama et al.
Journal of cellular physiology, 228(11), 2222-2231 (2013-04-30)
FUsed in Sarcoma/Translocated in LipoSarcoma (FUS/TLS or FUS) has been linked to several biological processes involving DNA and RNA processing, and has been associated with multiple diseases, including myxoid liposarcoma and amyotrophic lateral sclerosis (ALS). ALS-associated mutations cause FUS to
Shahram Attarian et al.
Orphanet journal of rare diseases, 9, 199-199 (2014-12-19)
Charcot-Marie-Tooth type 1A disease (CMT1A) is a rare orphan inherited neuropathy caused by an autosomal dominant duplication of a gene encoding for the structural myelin protein PMP22, which induces abnormal Schwann cell differentiation and dysmyelination, eventually leading to axonal suffering
J M van Griensven et al.
Clinical pharmacology and therapeutics, 58(6), 631-640 (1995-12-01)
To examine the effect of diabetes mellitus on the pharmacokinetics of tolrestat and to investigate its effect on red blood cell sorbitol levels according to a new pharmacodynamic model for this class of drugs. Single and multiple doses of tolrestat
Anna Boccaccio et al.
Cellular and molecular life sciences : CMLS, 71(21), 4275-4283 (2014-04-29)
Two-pore channel proteins (TPC) encode intracellular ion channels in both animals and plants. In mammalian cells, the two isoforms (TPC1 and TPC2) localize to the endo-lysosomal compartment, whereas the plant TPC1 protein is targeted to the membrane surrounding the large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service