Merck
Quantitative Validation of the
Chemosensory perception (2014-06-03)
Melanie Y Denzer, Stefan Gailer, David W Kern, L Philip Schumm, Norbert Thuerauf, Johannes Kornhuber, Andrea Buettner, Jonathan Beauchamp
ABSTRACT
Odorant pens are used by medical practitioners and researchers to assess olfactory dysfunction. Despite their routine use, there are currently no data on the gas-phase odorant concentrations released from the pen tips or whether these concentrations scale linearly with the aqueous-phase concentrations inside the pens. The commercially available Sniffin' Sticks odor threshold test containing
Product Description
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG