Merck
  • Quantification of bindings of organometallic ruthenium complexes to GSTπ by mass spectrometry.
Quantification of bindings of organometallic ruthenium complexes to GSTπ by mass spectrometry.
Journal of inorganic biochemistry (2015-03-15)
Yu Lin, Yongdong Huang, Wei Zheng, Kui Wu, Qun Luo, Yao Zhao, Shaoxiang Xiong, Fuyi Wang
ABSTRACT
Electrospray ionization mass spectrometry (ESI-MS) has been widely used to identify binding sites of metal complexes to proteins. However, the MS quantification of the metal-protein coordination remains a challenge. We have recently demonstrated by ESI-MS analysis that organometallic ruthenium complexes [(η(6)-arene)Ru(en)Cl](+) (arene=p-cymene (1), biphenyl (2) or 9,10-dihydrophenanthrene (3); en=ethylenediamine) bound to human glutathione-S-transferase π (GSTπ) at Cys15 and Cys48 within the G-site, and Cys102 and Met92 on the interface of the GSTπ dimer, showing inhibitory potency against the enzyme (J. Inorg. Biochem., 128 (2013) 77-84). Herein, we developed a mass spectrometric method to quantify the binding stoichiometry of the three complexes to GSTπ. The differences in signal intensities of the heavy-labelled peptides produced by tryptic digestion of the ruthenated GSTπ complexes and the respective light-labelled peptides in the tryptic digest of equimolar GSTπ were used to calculate the binding stoichiometry at specific residues. The results indicated that the pre-complexation of GSTπ with its substrate GSH significantly reduced the bindings of the ruthenium complexes at Met92 and Cys102, but had little impact on the bindings at Cys15 and Cys48. As the inhibitory activities of the ruthenium complexes against GSTπ are similar to those against GSTπ in complexation with GSH, these results suggest that the inhibition of the ruthenium complexes on GSTπ is attributed to the ruthenation at Cys15 and Cys48. The present work provides not only insights into the understanding on the inhibitory mechanism of ruthenium complexes GSTπ, but also a general method for quantitative characterization of metal-protein interactions.
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Acetoxy-d3-succinimide, 98 atom % D
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder