Merck
  • Determination of 15 sedative residues in mutton by rapid resolution liquid chromatography-tandem mass spectrometry.
Determination of 15 sedative residues in mutton by rapid resolution liquid chromatography-tandem mass spectrometry.
Journal of the science of food and agriculture (2014-06-21)
Jinmei Wei, Yuzhu Luo, Li Zhang, Suli Fang
ABSTRACT
The use of xenobiotic compounds in animal husbandry has given rise to consumer anxieties regarding residual risk and food safety. Thus, animal tissues have become main samples for residue analysis and food safety for sedatives. In this study, a rapid resolution liquid chromatography-tandem mass spectrometry (RRLC-MS/MS) method was established for the determination of 15 sedatives residues in mutton. After enzymolysis, sedatives residues in mutton were extracted by ammonium hydroxide-acetonitrile (10:90, v/v) and determined by RRLC-MS/MS with quantification by standard curve method. The calibration curves showed good linearity within the concentrations of 0.5-50 µg kg(-1) with the correlation coefficients (r(2)) ranged from 0.9639 to 0.9984. The limits of detection (LODs) and quantification (LOQs) were 0.25-2.5 and 0.5-5 µg kg(-1), respectively. The average recoveries of spikes samples were in the ranges of 74.1-116.8% with relative standard deviations of intra- and inter-day ranged from 2.6% to 11.2% and from 2.1% to 11.4%, respectively. This method is simple, sensitive and accurate in the determination of sedative residues.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Carazolol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Supelco
Methanol, analytical standard
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Xylazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)