Merck
  • Simultaneous determination of seventeen mycotoxins residues in Puerariae lobatae radix by liquid chromatography-tandem mass spectrometry.
Simultaneous determination of seventeen mycotoxins residues in Puerariae lobatae radix by liquid chromatography-tandem mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-06-18)
Shufang Wang, Ling Cheng, Shen Ji, Ke Wang
ABSTRACT
This work reported an efficient and accurate liquid chromatography tandem mass spectrometry (LC-MS/MS) method for simultaneous determination of seventeen mycotoxins in Puerariae lobatae radix, a frequently used traditional Chinese medicine (TCM). The effects of four different clean-up methods, including TC-M160, TC-T220, Mycosep 227, and QuEChERS method, on the recoveries of mycotoxins were investigated and compared. Finally, TC-M160 was chosen for better recovery and repeatability for mycotoxins analysis. The analytes were separated on an Agilent ZORBAX SB C18 column (4.6mm×250mm, 5μm particle size), and eluted with a mobile phase consisting of (A) water containing 0.1% formic acid and (B) acetonitrile containing 0.1% formic acid at a flow rate of 0.6mL/min. The separated compounds were detected by a triple quadrupole mass spectrometer operating in positive electrospray ionization with multiple reaction monitoring (MRM) mode. The results of method validation accorded with the requirement of analytical method for mycotoxins in COMMISSION REGULATION (EC) No 401/2006. The developed method was successfully applied for determination of mycotoxins in seventeen batches of Puerariae lobatae radix collected from different provinces of China. Three batches of them were found with contamination of mycotoxins AFB1 at (0.751±0.176)μg/kg, T-2 at (1.10±0.01)μg/kg, and T-2 at (0.853±0.044)μg/kg, respectively. The results demonstrated that the proposed method was suitable for monitoring mycotoxins residues in Puerariae lobatae radix.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
HT-2 Toxin solution, 100 μg/mL in acetonitrile, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Zearalanone
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Zearalenone, fungal mycotoxin
Sigma-Aldrich
HT-2 Toxin, ≥98% (HPLC)
Sigma-Aldrich
Alternariol monomethyl ether from Alternaria alternata (tenuis)
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%