Merck
  • A critical role for the long non-coding RNA GAS5 in proliferation and apoptosis in non-small-cell lung cancer.
A critical role for the long non-coding RNA GAS5 in proliferation and apoptosis in non-small-cell lung cancer.
Molecular carcinogenesis (2013-12-21)
Xuefei Shi, Ming Sun, Hongbing Liu, Yanwen Yao, Rong Kong, Fangfang Chen, Yong Song
ABSTRACT
In more recent years, long non-coding RNAs (lncRNAs) have been investigated as a new class of regulators of cellular processes, such as cell growth, apoptosis, and carcinogenesis. Although lncRNAs are dysregulated in numerous cancer types, limited data are available on the expression profile and functional role of lncRNAs in non-small cell lung cancer (NSCLC). In the present study, we determined the expression pattern of the growth arrest-specific transcript 5 (GAS5) in 72 NSCLC specimens by qRT-PCR and assess its biological functions in the development and progression of NSCLC. The results revealed that GAS5 expression was down-regulated in cancerous tissues compared to adjacent noncancerous tissues (P < 0.05) and was highly related to tumor size and TNM stage (P < 0.05). This correlation between GAS5 and clinicopathological parameters indicates that GAS5 might function as a tumor suppressor. Furthermore, GAS5 overexpression increased tumor cell growth arrest and induced apoptosis in vitro and in vivo. Meanwhile, siRNA-mediated knockdown of GAS5 promoted tumor cell growth. Importantly, through western blot analysis, we found that ectopic expression of GAS5 significantly up-regulated p53 expression and down-regulated transcription factor E2F1 expression. Taken together, these findings suggest that GAS5 is a tumor suppressor in NSCLC, and the action of GAS5 is mediated by p53-dependent and p53-independent pathways. GAS5 could serve as a potential diagnostic marker for NSCLC and may be a novel therapeutic target in patients with NSCLC.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Suz12
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ezh2
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting human SUZ12
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Monoclonal Anti-HLA Class I Antigen antibody produced in mouse, clone W6/32, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))