Merck
  • Siegesbeckia glabrescens attenuates allergic airway inflammation in LPS-stimulated RAW 264.7 cells and OVA induced asthma murine model.
Siegesbeckia glabrescens attenuates allergic airway inflammation in LPS-stimulated RAW 264.7 cells and OVA induced asthma murine model.
International immunopharmacology (2014-07-30)
Chan-Mi Jeon, In-Sik Shin, Na-Rae Shin, Ju-Mi Hong, Ok-Kyoung Kwon, Hui-Seong Kim, Sei-Ryang Oh, Pyung-Keun Myung, Kyung-Seop Ahn
ABSTRACT
Siegesbeckia glabrescens (SG) is a plant growing in Korea that is used as a traditional medicine for various inflammatory diseases. In this study, we investigated the protective effects of SG extract on allergic asthma in an ovalbumin (OVA)-induced asthma murine model and lipopolysaccharide (LPS)-stimulated RAW264.7 cells. Female BALB/c mice were sensitized by intraperitoneal injection of OVA on days 0 and 14 and then challenged with OVA from days 21 to 23. SG (30mg/kg) was administered by oral gavage 1h before the OVA challenge. LPS-stimulated RAW264.7 cells were evaluated to determine their levels of nitric oxide (NO). The SG significantly reduced the number of inflammatory cells in bronchoalveolar lavage (BAL) fluid and also reduced IL-4, IL-5, IL-13, eotaxin and immunoglobulin E in OVA-sensitized/challenged mice. SG also effectively reduced airway inflammation and mucus overproduction in lung tissue in addition to decreasing the expression of iNOS and COX-2. In LPS-stimulated RAW264.7 cells, SG treatment significantly reduced the levels of NO. These findings indicate that SG effectively suppressed inflammatory responses, and its effects appear to be related to reduction in iNOS and COX-2 expression. Therefore, we suggest that SG may have potential use as a therapeutic agent for inflammatory diseases such as allergic asthma.
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 60 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Sigma-Aldrich
Mouse IgE ELISA Kit
Sigma-Aldrich
Human Eotaxin / CCL11 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse Eotaxin ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin