Merck
  • Characterization of phosphorylation- and RNA-dependent UPF1 interactors by quantitative proteomics.
Characterization of phosphorylation- and RNA-dependent UPF1 interactors by quantitative proteomics.
Journal of proteome research (2014-04-26)
Valentin Flury, Umberto Restuccia, Angela Bachi, Oliver Mühlemann
ABSTRACT
Human up-frameshift 1 (UPF1) is an ATP-dependent RNA helicase and phosphoprotein implicated in several biological processes but is best known for its key function in nonsense-mediated mRNA decay (NMD). Here we employed a combination of stable isotope labeling of amino acids in cell culture experiments to determine by quantitative proteomics UPF1 interactors. We used this approach to distinguish between RNA-mediated and protein-mediated UPF1 interactors and to determine proteins that preferentially bind the hypo- or the hyper-phosphorylated form of UPF1. Confirming and expanding previous studies, we identified the eukaryotic initiation factor 3 (eIF3) as a prominent protein-mediated interactor of UPF1. However, unlike previously reported, eIF3 binds to UPF1 independently of UPF1's phosphorylation state. Furthermore, our data revealed many nucleus-associated RNA-binding proteins that preferentially associate with hyper-phosphorylated UPF1 in an RNase-sensitive manner, suggesting that UPF1 gets recruited to mRNA and becomes phosphorylated before being exported to the cytoplasm as part of the mRNP.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Anti-UPF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material