Merck
  • Developmental exposure to purity-controlled polychlorinated biphenyl congeners (PCB74 and PCB95) in rats: effects on brainstem auditory evoked potentials and catalepsy.
Developmental exposure to purity-controlled polychlorinated biphenyl congeners (PCB74 and PCB95) in rats: effects on brainstem auditory evoked potentials and catalepsy.
Toxicology (2014-12-03)
Hellmuth Lilienthal, Merja Korkalainen, Patrik L Andersson, Matti Viluksela
ABSTRACT
Whereas the effects of dioxin-like polychlorinated biphenyls (DL-PCBs) are well described, less is known about non-dioxin-like PCBs (NDL-PCBs), including influences on the nervous system and related behavioral effects after developmental exposure. Following the examination of the highly purified NDL congeners PCB52 and PCB180, we report here the results of experiments with PCB74 and PCB95. Rat dams were orally exposed to equimolar doses of either congener (40μmol/kg bw - 11.68mg PCB74/kg bw or 13.06mg PCB95/kg bw) from gestational day (GD) 10 to postnatal day (PND) 7. Control dams were given the vehicle. Adult offspring were tested for cataleptic behavior after induction with haloperidol, a classical neuroleptic drug, and brainstem auditory evoked potentials (BAEPs), using clicks and tone pips of different frequencies for stimulation. Results revealed slight effects on latencies to movement onset in female offspring exposed to PCB74, whereas PCB74 males and offspring exposed to PCB95 were not affected. Pronounced changes were observed in BAEPs at low frequencies in PCB74 offspring, with elevated thresholds in both sexes. PCB95 increased thresholds in males, but not females. Small effects were detected on latency of the late wave IV in both sexes after developmental exposure to PCB74 or PCB95. Compared with the other NDL-PCB congeners tested, PCB74 caused the most pronounced effects on BAEPs.
Sigma-Aldrich
Haloperidol, powder
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosgene solution, ~20% in toluene
Supelco
Haloperidol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosgene-12C solution, ~1 M in benzene, 99.9 atom % 12C
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%