Merck
  • Protective actions of aspirin-triggered (17R) resolvin D1 and its analogue, 17R-hydroxy-19-para-fluorophenoxy-resolvin D1 methyl ester, in C5a-dependent IgG immune complex-induced inflammation and lung injury.
Protective actions of aspirin-triggered (17R) resolvin D1 and its analogue, 17R-hydroxy-19-para-fluorophenoxy-resolvin D1 methyl ester, in C5a-dependent IgG immune complex-induced inflammation and lung injury.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-31)
Huifang Tang, Yanlan Liu, Chunguang Yan, Nicos A Petasis, Charles N Serhan, Hongwei Gao
ABSTRACT
Increasing evidence suggests that the novel anti-inflammatory and proresolving mediators such as the resolvins play an important role during inflammation. However, the functions of these lipid mediators in immune complex-induced lung injury remain unknown. In this study, we determined the role of aspirin-triggered resolvin D1 (AT-RvD1) and its metabolically stable analog, 17R-hydroxy-19-para-fluorophenoxy-resolvin D1 methyl ester (p-RvD1), in IgG immune complex-induced inflammatory responses in myeloid cells and injury in the lung. We show that lung vascular permeability in the AT-RvD1- or p-RvD1-treated mice was significantly reduced when compared with values in mice receiving control vesicle during the injury. Furthermore, i.v. administration of either AT-RvD1 or p-RvD1 caused significant decreases in the bronchoalveolar lavage fluid contents of neutrophils, inflammatory cytokines, and chemokines. Of interest, AT-RvD1 or p-RvD1 significantly reduced bronchoalveolar lavage fluid complement C5a level. By EMSA, we demonstrate that IgG immune complex-induced activation of NF-κB and C/EBPβ transcription factors in the lung was significantly inhibited by AT-RvD1 and p-RvD1. Moreover, AT-RvD1 dramatically mitigates IgG immune complex-induced NF-κB and C/EBP activity in alveolar macrophages. Also, secretion of TNF-α, IL-6, keratinocyte cell-derived chemokine, and MIP-1α from IgG immune complex-stimulated alveolar macrophages or neutrophils was significantly decreased by AT-RvD1. These results suggest a new approach to the blocking of immune complex-induced inflammation.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Mouse C5a / Complement C5a ELISA Kit, for serum, plasma and cell culture supernatants
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Human C5a / Complement C5a ELISA Kit, for serum, plasma, and cell culture supernatants
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Supelco
cis-4,7,10,13,16,19-Docosahexaenoic acid, analytical standard