Merck
  • Stability and removal of dexamethasone sodium phosphate from wastewater using modified clays.
Stability and removal of dexamethasone sodium phosphate from wastewater using modified clays.
Environmental technology (2014-06-25)
Saleh Sulaimana, Mustafa Khamis, Shlomo Nir, Filomena Lelario, Laura Scrano, Sabino Aurelio Bufo, Rafik Karaman
ABSTRACT
Stability and removal of dexamethasone sodium phosphate (DSP) from wastewater produced at Al-Quds University Campus were investigated. Kinetic studies in both pure water and wastewater coming from secondary treatment (activated sludge) demonstrated that the anti-inflammatory DSP underwent degradation to its hydrolytic derivative, dexamethasone, in both media. The first-order hydrolysis rate of DSP in activated sludge at 25 degrees C (3.80 x 10(-6) s-1) was about 12-fold larger than in pure water (3.25 x 10(-7) s-1). The overall performance of the wastewater treatment plant (WWTP) installed in the University Campus was also assessed showing that 90% of spiked DSP was removed together with its newly identified metabolites by the ultra-filtration (UF) system, which consists of a UF hollow fibre (HF) with a 100-kDa cutoff membrane as the pre-polishing stage for the UF spiral wound with a 20-kDa cutoffmembrane. In testing other technologies, the effectiveness of adsorption and filtration by micelle-clay (MC) preparation for removing DSP was ascertained in comparison with activated charcoal. Batch adsorption in aqueous suspensions of the MC composite and activated carbon was well described by Langmuir isotherms showing the best results for MC material. Filtration of DSP water solutions demonstrated a significant advantage of columns filled in with a mixture of sand and MC complex in comparison with activated carbon/sand filters.
Sigma-Aldrich
Dexamethasone 21-phosphate disodium salt, ≥98%
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone sodium phosphate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone sodium phosphate for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard