Merck
  • Metabolic reengineering invoked by microbial systems to decontaminate aluminum: implications for bioremediation technologies.
Metabolic reengineering invoked by microbial systems to decontaminate aluminum: implications for bioremediation technologies.
Biotechnology advances (2012-12-04)
Christopher Auger, Sungwon Han, Varun P Appanna, Sean C Thomas, Gerardo Ulibarri, Vasu D Appanna
ABSTRACT
As our reliance on aluminum (Al) increases, so too does its presence in the environment and living systems. Although generally recognized as safe, its interactions with most living systems have been nefarious. This review presents an overview of the noxious effects of Al and how a subset of microbes can rework their metabolic pathways in order to survive an Al-contaminated environment. For instance, in order to expulse the metal as an insoluble precipitate, Pseudomonas fluorescens shuttles metabolites toward the production of organic acids and lipids that play key roles in chelating, immobilizing and exuding Al. Further, the reconfiguration of metabolic modules enables the microorganism to combat the dearth of iron (Fe) and the excess of reactive oxygen species (ROS) promoted by Al toxicity. While in Rhizobium spp., exopolysaccharides have been invoked to sequester this metal, an ATPase is known to safeguard Anoxybacillus gonensis against the trivalent metal. Hydroxyl, carboxyl and phosphate moieties have also been exploited by microbes to trap Al. Hence, an understanding of the metabolic networks that are operative in microorganisms residing in polluted environments is critical in devising bioremediation technologies aimed at managing metal wastes. Metabolic engineering is essential in elaborating effective biotechnological processes to decontaminate metal-polluted surroundings.
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 6.0mm, as drawn, 99.999%
Aluminum, microfoil, disks, 50mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, rod, 44mm, diameter 77.5mm, as drawn, 99.999%
Aluminum, tube, 200mm, outside diameter 0.86mm, inside diameter 0.76mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, microfoil, disks, 25mm, thinness 0.0075μm, specific density 1.84μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, wire reel, 100m, diameter 0.5mm, as drawn, 99.95+%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, tube, 100mm, outside diameter 0.6mm, inside diameter 0.3mm, wall thickness 0.15mm, as drawn, 99.5%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, tube, 200mm, outside diameter 1.42mm, inside diameter 1.166mm, wall thickness 0.127mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 10.0mm, as drawn, 99.999%
Aluminum, tube, 500mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, rod, 200mm, diameter 12.7mm, 99%
Aluminum, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%