Skip to Content
Merck
All Photos(1)

Key Documents

IRMM523B

Aluminum

IRMM®, certified reference material, 1.0 mm foil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

form

foil

autoignition temp.

1400 °F

manufacturer/tradename

JRC

resistivity

2.6548 μΩ-cm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Analysis Note

For more information please see:
IRMM523B

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Al18F: a new standard for radiofluorination.
David M Goldenberg et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(7), 1170-1170 (2013-05-21)
Seong-Hee Lee et al.
Journal of nanoscience and nanotechnology, 13(1), 509-512 (2013-05-08)
An ultrafine grain (UFG) complex lamella aluminum alloy sheet was successfully fabricated by ARB process using AA1050 and AA6061. The lamella thickness of the alloy became thinner and elongated to the rolling direction with increasing the number of ARB cycles.
Kate M Brougham et al.
Environmental pollution (Barking, Essex : 1987), 178, 89-96 (2013-04-03)
Although a great deal is known about the deposition of fluoride on vegetation, and the hazards associated with uptake by grazing herbivores, little is known about what happens to the concentration of fluoride in vegetation and soil at polluted sites
Chih-Hung Guo et al.
Nutrients, 5(4), 1456-1470 (2013-04-24)
End stage renal disease patients undergoing long-term dialysis are at risk for abnormal concentrations of certain essential and non-essential trace metals and high oxidative stress. We evaluated the effects of zinc (Zn) supplementation on plasma aluminum (Al) and selenium (Se)
A S Waters et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(8), 1764-1772 (2013-04-13)
Acid mine drainage (AMD) has degraded water quality and ecology in streams on the Stockton Plateau, the site of New Zealand's largest open-cast coal mining operation. This has previously been attributed largely to the effects of acidity and elevated aluminium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service