Merck
  • Thalassospira povalilytica sp. nov., a polyvinyl-alcohol-degrading marine bacterium.
Thalassospira povalilytica sp. nov., a polyvinyl-alcohol-degrading marine bacterium.
International journal of systematic and evolutionary microbiology (2014-01-11)
Yuichi Nogi, Masaki Yoshizumi, Masayuki Miyazaki
ABSTRACT
A polyvinyl-alcohol-degrading marine bacterium was isolated from plastic rope litter found in Tokyo Bay, Japan. The isolated strain, Zumi 95(T), was a Gram-reaction-negative, non-spore-forming and facultatively anaerobic chemo-organotroph. The major respiratory quinone was Q-10. The predominant fatty acids were C18 : 1ω7c and C16 : 0. On the basis of 16S rRNA gene sequence analysis, the isolated strain was closely affiliated with members of the genus Thalassospira in the class Alphaproteobacteria. The DNA G+C content of the novel strain was 55.1 mol%. The hybridization values for DNA-DNA relatedness between this strain and four reference strains representing species of the genus Thalassospira were significantly lower than that accepted as the phylogenetic definition of a species. On the basis of differences in taxonomic characteristics, the isolated strain represents a novel species of the genus Thalassospira for which the name Thalassospira povalilytica sp. nov. (type strain Zumi 95(T) = JCM 18746(T) = DSM 26719(T)) is proposed.
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Coenzyme Q2, ≥90%
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Poly(vinyl alcohol), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000