Skip to Content
Merck
All Photos(2)

Key Documents

363146

Sigma-Aldrich

Poly(vinyl alcohol)

Mw 85,000-124,000, 99+% hydrolyzed

Synonym(s):

PVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

Quality Level

form

powder or crystals

mol wt

Mw 85,000-124,000

falling ball

28-32 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly (vinyl alcohol) (PVA) is a useful industrial, medical and biomimetic material. PVA with high molecular weight is useful in preparing gel which possesses both high strength and modulus. Hence, it is used for the production of fibers, films or gels.

Application

  • Poly (vinyl alcohol) recent contributions to engineering and medicine: This article discusses the diverse applications of PVA in engineering and medical fields, emphasizing its versatility and potential in various domains (Feldman, 2020).
  • Chemical modification of poly (vinyl alcohol) in water: This study explores the chemical modification processes of PVA, enhancing its functional properties for advanced material applications (Awada & Daneault, 2015).
  • Crosslinked poly (vinyl alcohol) hydrogels for wound dressing applications: Reviews the use of PVA in creating hydrogel dressings for medical applications, demonstrating the material′s effectiveness in healing and treatment scenarios (Kamoun et al., 2015).
  • A review on mechanical and water absorption properties of polyvinyl alcohol based composites/films: This review focuses on the mechanical and water absorption properties of PVA composites, crucial for developing advanced materials in various scientific and industrial applications (Jain et al., 2017).
  • Properties and applications of polyvinyl alcohol, halloysite nanotubes and their nanocomposites: Analyzes the synthesis, properties, and applications of PVA-based nanocomposites, highlighting their significant potential in nanotechnology and material science (Gaaz et al., 2015).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Active chitosan/PVA films with anthocyanins from Brassica oleraceae (Red Cabbage) as Time-Temperature Indicators for application in intelligent food packaging
Pereira VA Jr, et al.
Food Hydrocolloids, 43, 180-188 (2015)
B D Price et al.
Physics in medicine and biology, 55(4), 1177-1188 (2010-01-29)
We have developed a novel phantom material: a solution of polyvinyl alcohol (PVAL) in ethanol and water, freeze-thawed to produce a solid yet elastically compressible gel. The x-ray attenuation and mechanical properties of these gels are compared with published measurements
Design and development of papain-urea loaded PVA nanofibers for wound debridement
Shoba E, et al.
Royal Society of Chemistry Advances, 4(104), 60209-60215 (2014)
Synthesis and characterization of CdS quantum dots with carboxylic-functionalized poly (vinyl alcohol) for bioconjugation
Mansur HS, et al.
Polymer, 52(4), 1045-1054 (2011)
Hua Yang et al.
Molecular therapy. Nucleic acids, 20, 451-458 (2020-04-11)
We have reported that of the 10 most commonly used adeno-associated virus (AAV) serotype vectors, AAV6 is the most efficient in transducing primary human hematopoietic stem cells (HSCs) in vitro, as well as in vivo. More recently, polyvinyl alcohol (PVA), was reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service