Merck
  • Performance evaluation of non-targeted peak-based cross-sample analysis for comprehensive two-dimensional gas chromatography-mass spectrometry data and application to processed hazelnut profiling.
Performance evaluation of non-targeted peak-based cross-sample analysis for comprehensive two-dimensional gas chromatography-mass spectrometry data and application to processed hazelnut profiling.
Journal of chromatography. A (2012-05-11)
Johannes Kiefl, Chiara Cordero, Luca Nicolotti, Peter Schieberle, Stephen E Reichenbach, Carlo Bicchi
ABSTRACT
The continuous interest in non-targeted profiling induced the development of tools for automated cross-sample analysis. Such tools were found to be selective or not comprehensive thus delivering a biased view on the qualitative/quantitative peak distribution across 2D sample chromatograms. Therefore, the performance of non-targeted approaches needs to be critically evaluated. This study focused on the development of a validation procedure for non-targeted, peak-based, GC×GC-MS data profiling. The procedure introduced performance parameters such as specificity, precision, accuracy, and uncertainty for a profiling method known as Comprehensive Template Matching. The performance was assessed by applying a three-week validation protocol based on CITAC/EURACHEM guidelines. Optimized ¹D and ²D retention times search windows, MS match factor threshold, detection threshold, and template threshold were evolved from two training sets by a semi-automated learning process. The effectiveness of proposed settings to consistently match 2D peak patterns was established by evaluating the rate of mismatched peaks and was expressed in terms of results accuracy. The study utilized 23 different 2D peak patterns providing the chemical fingerprints of raw and roasted hazelnuts (Corylus avellana L.) from different geographical origins, of diverse varieties and different roasting degrees. The validation results show that non-targeted peak-based profiling can be reliable with error rates lower than 10% independent of the degree of analytical variance. The optimized Comprehensive Template Matching procedure was employed to study hazelnut roasting profiles and in particular to find marker compounds strongly dependent on the thermal treatment, and to establish the correlation of potential marker compounds to geographical origin and variety/cultivar and finally to reveal the characteristic release of aroma active compounds.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler