Merck
  • Extraction of carotenoids from feces, enabling the bioavailability of beta-carotene to be studied in Indonesian children.
Extraction of carotenoids from feces, enabling the bioavailability of beta-carotene to be studied in Indonesian children.
Journal of agricultural and food chemistry (2003-08-09)
Machteld Van Lieshout, Clive E West, Peter Van De Bovenkamp, Yan Wang, Yongkai Sun, Richard B Van Breemen, Dewi Permaesih Muhilal, Michiel A Verhoeven, Alain F L Creemers, Johan Lugtenburg
ABSTRACT
Previously, we have presented a method for quantifying beta-carotene bioavailability based on analysis in serum, following administration of (13)C-labeled beta-carotene. Because stool samples can be collected noninvasively, we have now extended the method to measure the bioavailability based on measurements in feces. An extraction method was developed to enable measurement of concentrations and degree of isotopic enrichment of retinol, retinyl palmitate and carotenoids in feces. Relative bioavailability of beta-carotene from pumpkin (n = 6) was found to be 1.8 times (interval, 0.6, 5.5) greater than that from spinach (n = 8), based on data from feces compared with 1.7 times (interval, 0.9, 3.1) based on data from serum.
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Supelco
Retinol palmitate, analytical standard
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material