Merck
Ethinylestradiol/Chlormadinone acetate: dermatological benefits.
American journal of clinical dermatology (2011-09-16)
Aurora Guerra-Tapia, Blanca Sancho Pérez
ABSTRACT
Acne vulgaris, hirsutism, seborrhea and female pattern hair loss (FPHL) are common disorders of the pilosebaceous unit (PSU). In some women with hyperandrogenemia, an excess of androgens at the PSU can lead to the development of these dermatological manifestations. These manifestations can cause many psychiatric and psychological implications, such as social fears and anxiety, and can adversely affect quality of life. High androgen levels at the PSU as a possible underlying cause of acne vulgaris, hirsutism, seborrhea and FPHL supports the rationale for using combined oral contraceptives for the management of these conditions in women. The purpose of this review is to describe these dermatological manifestations of the PSU and the management of these conditions through the use of the oral contraceptive ethinylestradiol/chlormadinone acetate (EE/CMA). EE/CMA 0.03/2 mg is a combined monophasic contraceptive pill with anti-androgenic properties. It is approved in Europe for contraception and has been investigated in phase III trials for the treatment of acne. EE/CMA was better than placebo and similar to another low-dose oral contraceptive (ethinylestradiol/levonorgestrel) in improving symptoms of acne in two phase III randomized controlled trials in patients with mild to moderate papulopustular acne. In addition, in trials investigating the contraceptive efficacy of EE/CMA, limited data suggest that there were also improvements in hirsutism, FPHL and seborrhea in small subgroups of patients. EE/CMA has a good safety profile. The most commonly reported adverse events are breast tenderness/pain, headache/migraine and nausea. Evidence in the literature indicates that the use of EE/CMA for the treatment of dermatological disorders under the control of androgens may be a valid treatment option. Further investigation is warranted.
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chlormadinone acetate, ≥98%
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
Chlormadinone acetate, European Pharmacopoeia (EP) Reference Standard