Skip to Content
Merck
All Photos(1)

Key Documents

Y0001124

Ethinylestradiol for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

17α-Ethynylestradiol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
2419975
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ethinylestradiol

manufacturer/tradename

EDQM

mp

182-183 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@@]3(O)C#C)[C@]1([H])CCc4cc(O)ccc24

InChI

1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1

InChI key

BFPYWIDHMRZLRN-SLHNCBLASA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethinylestradiol for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jürgen Dinger et al.
Obstetrics and gynecology, 122(4), 800-808 (2013-10-03)
To compare the risks of short-term and long-term use of an etonogestrel-containing and ethinylestradiol-containing vaginal ring and combined oral contraceptive pills (OCPs) in a routine clinical study population. This was a prospective, controlled, noninterventional cohort study performed in the United
Bin Huang et al.
Environment international, 59, 262-273 (2013-07-16)
The occurrence, removal and bioaccumulation of steroid estrogens such as the natural estrone (E1), 17β-estradiol (E2) and estriol (E3), as well as the synthetic 17α-ethynylestradiol (EE2) were investigated in Dianchi Lake catchment, China. The results show that traditional secondary treatment
A Frølich et al.
Bone, 11(6), 393-396 (1990-01-01)
An attempt was made to explain the bone-preserving effect of estrogen by analysis of estrogen receptors (OER) in the calcitonin-producing C-cells of the human thyroid gland. Thyroid tissue from twenty patients with benign hyperthyroidism and three patients with medullary thyroid
Konrad F Koehler et al.
Endocrine reviews, 26(3), 465-478 (2005-04-29)
We have known for many years that estrogen is more than the female hormone. It is essential in the male gonads, and in both sexes, estrogen has functions in the skeleton and central nervous system, on behavior, and in the
Jie Han et al.
Water research, 47(7), 2273-2284 (2013-03-08)
This study demonstrates that ethinylestradiol (EE2), a priority estrogenic contaminant in water, can be rapidly and selectively removed from aqueous solutions using industrial-grade polyamide 612 (PA612) particles as adsorbents. Isothermal studies showed that nonporous low surface area (20 m(2) g(-1)) PA612 particles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service