Merck
  • Monitoring of exposure to methylpentanes by diffusive sampling and urine analysis for alcoholic metabolites.
Monitoring of exposure to methylpentanes by diffusive sampling and urine analysis for alcoholic metabolites.
Occupational and environmental medicine (1995-11-01)
T Kawai, K Mizunuma, T Yasugi, S Horiguchi, H Iguchi, A Mutti, S Ghittori, M Ikeda
ABSTRACT
To investigate the possibilities of personal ambient monitoring and biological monitoring for methylpentane isomers. The performance of activated carbon cloth to absorb 2- and 3-methylpentane was studied by experimental vapour exposure followed by solvent extraction and gas chromatography (GC). Urine from workers and rats exposed to 2- and 3-methylpentane was analysed by GC with or without acid or enzymatic hydrolysis. Carbon cloth absorbed 2- and 3-methylpentane linearly to exposures up to eight hours and to 400 ppm, and was sensitive enough to detect a 15 minute peak of exposure. The two isomers were clearly separated from hexane on a DB-1 column. For analysis of the urine, enzymatic hydrolysis was superior to acid hydrolysis. Exposure of rats to methylpentane vapours showed that 2-methyl-2-pentanol and 3-methyl-2-pentanol were excreted in urine in proportion to the dose of 2-methylpentane and 3-methylpentane, respectively. 2-Methyl derivatives of 1-, 3-, and 4-propanol, 2-methylpentane-2,4-diol, and 3-methyl-2-pentanol were minor metabolites. Analysis of urine from the exposed workers showed that 2-methyl- and 3-methyl-2-pentanol are leading urinary metabolites after exposure to the corresponding methylpentane. Diffusive sampling is applicable to monitor 2- and 3-methylpentane vapours as is the case for hexane vapour. 2-Methyl-2-pentanol and 3-methyl-2-pentanol will be markers of occupational exposure to 2-methylpentane and 3-methylpentane, respectively. Also, 2-methylpentane-2,4-diol might be a marker of exposure to 2-methylpentane.
Sigma-Aldrich
3-Methylpentane, ≥99%
Supelco
3-Methylpentane, analytical standard
Sigma-Aldrich
2-Methyl-2-pentanol, 99%