Merck
  • The pulmonary capillary blood flow (Qc) at rest and work measured simultaneously by 3 inert substances.
The pulmonary capillary blood flow (Qc) at rest and work measured simultaneously by 3 inert substances.
Giornale italiano di medicina del lavoro (1993-09-01)
M Imbriani
ABSTRACT
We have considered the opportunity of using some inert vapours (2-3 Dimethylbutane, 2 Methylpentane, n-Hexane) to measure the pulmonary capillary blood flow (Qc). The measurements were performed by means of a gas chromatograph connected with a mass spectrometer. The alpha values (Bunsen absorption coefficients) measured by a constant pressure method (multiple equilibration methods) were reported. Qc was measured in 14 healthy subjects at rest and in 5 subjects during exercise (25 and 50 watt by means of a cyclo ergometer, steady state test) (rebreathing method). The method used allow multiple measurements of Qc during the same exercise test.
Sigma-Aldrich
2,3-Dimethylbutane, 98%
Sigma-Aldrich
2-Methylpentane, ≥99%
Sigma-Aldrich
2-Methylpentane, ≥95.0% (GC)
Supelco
2-Methylpentane, analytical standard