Skip to Content
End of year dates: Orders must be billed by 05/12. Merck Mexico will be closed from 18/12 to 02/01.
Merck
  • Oro-facial granulomatosis. Response to elemental diet and provocation by food additives.
Oro-facial granulomatosis. Response to elemental diet and provocation by food additives.
Clinical allergy (1986-07-01)
M C Sweatman, R Tasker, J O Warner, M M Ferguson, D N Mitchell
ABSTRACT
We report the case of an 8.5-year-old girl with oro-facial granulomatosis associated with clinical atopy, in whom relapse of her granulomatous disorder was shown to be related to exposure to specific food additives, viz. carmoisine, sunset yellow and monosodium glutamate. Treatment with a restricted diet resulted in considerable regression in the facial swelling which has been maintained for 6 months. A brief account of the histological features, both under light and electron microscopy, is given, together with a description of the use of nuclear magnetic resonance scanning in the assessment of this disease. The patient had no evidence to support a diagnosis of sarcoidosis or Crohn's disease.
Product Description
Supelco
Carmoisine, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Chromotrope FB, Dye content 50 %
Pricing and availability is not currently available.