Merck
  • Synthesis of new quinolinequinone derivatives and preliminary exploration of their cytotoxic properties.
Synthesis of new quinolinequinone derivatives and preliminary exploration of their cytotoxic properties.
Journal of medicinal chemistry (2013-04-12)
Charles M Keyari, Alison K Kearns, Nathan S Duncan, Emily A Eickholt, Geoffrey Abbott, Howard D Beall, Philippe Diaz
ABSTRACT
A series of 7-amino- and 7-acetamidoquinoline-5,8-diones with aryl substituents at the 2-position were synthesized, characterized, and evaluated as potential NAD(P)H:quinone oxidoreductase (NQO1) -directed antitumor agents. The synthesis of lavendamycin analogues is illustrated. Metabolism studies demonstrated that 7-amino analogues were generally better substrates for NQO1 than 7-amido analogues, as were compounds with smaller heteroaromatic substituents at the C-2 position. Surprisingly, only two compounds, 7-acetamido-2-(8'-quinolinyl)quinoline-5,8-dione (11) and 7-amino-2-(2-pyridinyl)quinoline-5,8-dione (23), showed selective cytotoxicity toward the NQO1-expressing MDA468-NQ16 breast cancer cells versus the NQO1-null MDA468-WT cells. For all other compounds, NQO1 protected against quinoline-5,8-dione cytotoxicity. Compound 22 showed potent activity against human breast cancer cells expressing or not expressing NQO1, with respective IC50 values of 190 nM and 140 nM and a low NQO1-mediated reduction rate, which suggests that the mode of action of 22 differs from that of lavendamycin and involves an unidentified target(s).
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% (GE)
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis