Merck
  • Hydroxytyrosyl ethyl ether exhibits stronger intestinal anticarcinogenic potency and effects on transcript profiles compared to hydroxytyrosol.
Hydroxytyrosyl ethyl ether exhibits stronger intestinal anticarcinogenic potency and effects on transcript profiles compared to hydroxytyrosol.
Food chemistry (2013-02-16)
G Pereira-Caro, R Mateos, M H Traka, J R Bacon, R Bongaerts, B Sarriá, L Bravo, P A Kroon
ABSTRACT
The anticarcinogenic activity of hydroxytyrosyl ethyl ether (HTy-Et) compared to its precursor hydroxytyrosol (HTy) has been studied in human Caco-2 colon adenocarcinoma cells. 451 and 977 genes were differentially expressed in Caco-2 cells exposed to HTy or HTy-Et for 24h, respectively, compared with untreated cells (P<0.005; FDR=0), using Affymetrix microarrays. Results showed that both HTy and HTy-Et inhibited cell proliferation and arrested the cell cycle by up-regulating p21 and CCNG2 and down-regulating CCNB1 protein expression. HTy and HTy-Et also altered the transcription of specific genes involved in apoptosis, as suggested by the up-regulation of BNIP3, BNIP3L, PDCD4 and ATF3 and the activation of caspase-3. Moreover, these polyphenols up-regulated xenobiotic metabolizing enzymes UGT1A10 and CYP1A1, enhancing carcinogen detoxification. In conclusion, these results highlight that HTy and its derivative HTy-Et modulate molecular mechanisms involved in colon cancer, with HTy-Et being more effective than HTy.
Product Description
Supelco
Phenylethyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
3-Hydroxytyrosol, ≥98% (HPLC)