Merck
GaN light emitting diodes with wing-type imbedded contacts.
Optics express (2013-02-15)
Ray-Hua Horng, Kun-Ching Shen, Yu-Wei Kuo, Dong-Sing Wuu
ABSTRACT
A wing-type imbedded electrodes was introduced into the lateral light emitting diode configuration (WTIE-LEDs) to reduce the effect of light shading of electrode in conventional sapphire-based LEDs (CSB-LEDs). The WTIE-LEDs with double-side roughened surface structures not only can eliminate the light shading of electrode and bonding wire, but also increase the light extraction and light output power. Contrast to CSB-LEDs, a 79% enhancement of output intensity in the WTIE-LED was obtained at 100 mA injection current. Similarly, the output power of packaged WTIE-LEDs was enhanced 59% higher compared with the packaged CSB-LEDs at the same injection condition. Therefore, using the imbedded contact to reduce light shading would be a promising prospective for LEDs to achieve high output power.
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, bars, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis