Merck
  • Methoxyethylammonium monosubstituted β-cyclodextrin as the chiral selector for enantioseparation in capillary electrophoresis.
Methoxyethylammonium monosubstituted β-cyclodextrin as the chiral selector for enantioseparation in capillary electrophoresis.
Journal of chromatography. A (2013-01-12)
Shuye Wang, Yun Dai, Jianhua Wu, Jie Zhou, Jian Tang, Weihua Tang
ABSTRACT
Methoxyethylamine monosubstituted β-cyclodextrin, mono-6(A)-(2-methoxyethyl-1-ammonium)-6(A)-β-cyclodextrin chloride (MEtAMCD), is synthesized and analytically characterized. Bearing a methoxy group in cyclodextrin rim, MEtAMCD exhibits outstanding enantioselectivities toward ampholytic and acidic racemates in capillary electrophoresis. Driven by inclusion complexation, electrostatic interactions and/or hydrogen bonding, the enantioseparation of MEtAMCD is found to be strongly dependent on various separation parameters including buffer pH, cyclodextrin concentration, applied voltage, separation temperature and organic solvent additives. MEtAMCD demonstrates as a versatile cationic chiral selector for the studied 26 acidic and ampholytic enantiomers.
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)