Merck
  • Copper nanoparticles mediated by chitosan: synthesis and characterization via chemical methods.
Copper nanoparticles mediated by chitosan: synthesis and characterization via chemical methods.
Molecules (Basel, Switzerland) (2012-12-18)
Muhammad Sani Usman, Nor Azowa Ibrahim, Kamyar Shameli, Norhazlin Zainuddin, Wan Md Zin Wan Yunus
ABSTRACT
Herein we report a synthesis of copper nanoparticles (Cu-NPs) in chitosan (Cts) media via a chemical reaction method. The nanoparticles were synthesized in an aqueous solution in the presence of Cts as stabilizer and CuSO(4)·5H(2)O precursor. The synthesis proceeded with addition of NaOH as pH moderator, ascorbic acid as antioxidant and hydrazine( )as the reducing agent. The characterization of the prepared NPs was done using ultraviolet-visible spectroscopy, which showed a 593 nm copper band. The Field Emission Scanning Electron Microscope (FESEM) images were also observed, and found to be in agreement with the UV-Vis result, confirming the formation of metallic Cu-NPs. The mean size of the Cu-NPs was estimated to be in the range of 35-75 nm using X-ray diffraction. XRD was also used in analysis of the crystal structure of the NPs. The interaction between the chitosan and the synthesized NPs was studied using Fourier transform infrared (FT-IR) spectroscopy, which showed the capping of the NPs by Cts.
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine, anhydrous, 98%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%