Merck
  • pH-indicators doped polysaccharide LbL coatings for hazardous gases optical sensing.
pH-indicators doped polysaccharide LbL coatings for hazardous gases optical sensing.
Carbohydrate polymers (2012-12-12)
A Yu Mironenko, A A Sergeev, S S Voznesenskiy, D V Marinin, S Yu Bratskaya
ABSTRACT
Sensitive layer-by-layer (LbL) coatings for optical detection of gaseous NH(3) and HCl were prepared by self-assembly of oppositely charged polysaccharides (chitosan and λ-carrageenan) followed by doping LbLs with pH-sensitive dyes - bromothymol blue (BTB) and Congo red (CR). It has been shown that CR, being an amphoteric dye, diffuses into LbL films regardless of the charge of the outermost polyelectrolyte layer, and the dye loading increases linearly with the LbL film thickness, whereas BTB diffuses into LbL films only when the outermost layer is positively charged, and linearity between dye loading and film thickness holds only up to 8-12 double layers (DLs) deposited. Formation of dye-doped LbL coatings at the surface of K(+)/Na(+) ion-exchanged glass has allowed fabrication of composite optical waveguide (OWG) gas sensor for detection of ammonia and hydrochloric acid vapors. The response time of BTB-doped composite OWG for ammonia detection was below 1s, and the detection limit was below 1 ppm. CR-doped OWG sensors have shown high sensitivity to HCl vapor but slow relaxation time (up to several hours for 12 DL LbL films).
Product Description
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Congo Red, BioXtra, certified by the Biological Stain Commission
Sigma-Aldrich
Congo Red, Dye content ≥35 %