Merck
  • Label-free aptasensor for thrombin determination based on the nanostructured phenazine mediator.
Label-free aptasensor for thrombin determination based on the nanostructured phenazine mediator.
Talanta (2012-11-28)
Gennady A Evtugyn, Veronika B Kostyleva, Anna V Porfireva, Maria A Savelieva, Vladimir G Evtugyn, Rusal R Sitdikov, Ivan I Stoikov, Igor S Antipin, Tibor Hianik
ABSTRACT
New aptasensors based on DNA aptamer and polycarboxylated thiacalix[4]arenes in cone, 1,3-alternate and partial cone configurations bearing Neutral Red (NR) at substituents at the lower rim have been developed and applied for thrombin detection. The assembly of the biorecognition layer was optimized by AFM and EIS study to reach the maximal coverage and regular composition of the surface layer. The interaction of the NR groups with thrombin suppressed the electron hopping between oxidized and reduced mediator groups. This regularly decreased the NR peak current and increased the resistance of the charge transfer. The aptasensor makes it possible to detect from 1 nM to 1 μM of thrombin with the detection limit of 0.05-0.5 nM. No effect of the 1000 excess of bovine serum albumin on the signal was observed. The influence of thiacalix[4]arene configuration on the sensitivity of aptasensor signal toward thrombin is discussed.
Sigma-Aldrich
Neutral Red, for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0)
Sigma-Aldrich
Neutral Red, certified by the Biological Stain Commission
Sigma-Aldrich
Neutral Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red Solution (0.33%), 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red, Dye content ≥90 %
Sigma-Aldrich
Neutral Red Solution, Buffered