Merck
  • Efficient photocatalytic reduction of aqueous Cr(VI) over flower-like SnIn4S8 microspheres under visible light illumination.
Efficient photocatalytic reduction of aqueous Cr(VI) over flower-like SnIn4S8 microspheres under visible light illumination.
Journal of hazardous materials (2012-11-28)
Lin Wang, Xinyong Li, Wei Teng, Qidong Zhao, Yong Shi, Renliang Yue, Yunfa Chen
ABSTRACT
Photocatalytic reduction of aqueous Cr(VI) was successfully achieved on nanostructured SnIn(4)S(8). The SnIn(4)S(8) particles with flower-like nanostructure were synthesized via a facile solvothermal method. UV-vis diffuse reflectance spectra (DRS) indicated that the SnIn(4)S(8) particles had strong absorption in visible region and the band gap was estimated to be from 2.27 to 2.35 eV. The photocatalytic reduction of aqueous Cr(VI) by flower-like SnIn(4)S(8) was evaluated under visible light (λ>400 nm) irradiation. The polyvinyl pyrrolidone (PVP) assisted SnIn(4)S(8) sample exhibits excellent removal efficiency of Cr(VI) (~97%) and good photocatalytic stability. The predominant photocatalytic activity is due to its large surface area, strong absorption in visible-light region and excellent charge separation characteristics.
Product Description
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%