Merck
  • DNA-based diagnostics for genetically modified cotton: decaplex PCR assay to differentiate MON531 and MON15985 Bt cotton events.
DNA-based diagnostics for genetically modified cotton: decaplex PCR assay to differentiate MON531 and MON15985 Bt cotton events.
Methods in molecular biology (Clifton, N.J.) (2012-11-13)
Gurinder Jit Randhawa, Monika Singh, Rashmi Chhabra
ABSTRACT
The adoption rate and global area under cultivation of genetically modified (GM) crops is dramatically increasing in recent past. GM cotton has occupied 25.0 million hectares (mha) comprising 15.6% of the global area under GM cultivation. Bt cotton, expressing delta-endotoxins from Bacillus thuringiensis (Bt), is the only commercialized crop in India that is planted on an area of 10.6 mha. With the increase in development and commercialization of GM crops, it is necessary to develop appropriate qualitative and quantitative methods for detection of different GM events. Robust diagnostics for GM detection need to be developed and implemented to monitor and detect different events of GM cotton in India. This chapter summarizes the methods based on polymerase chain reaction (PCR) being employed for detection of different GM events of cotton. We describe a decaplex PCR method for identification and differentiation of two major commercialized events of Bt cotton, i.e., MON531 and MON15985, in India.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%