Merck
  • Potassium iodide catalyzed simultaneous C3-formylation and N-aminomethylation of indoles with 4-substituted-N,N-dimethylanilines.
Potassium iodide catalyzed simultaneous C3-formylation and N-aminomethylation of indoles with 4-substituted-N,N-dimethylanilines.
Organic & biomolecular chemistry (2012-11-13)
Lan-Tao Li, Hong-Ying Li, Li-Juan Xing, Li-Juan Wen, Peng Wang, Bin Wang
ABSTRACT
A one-pot dual functionalization of indoles has been developed. The simultaneous C3-formylation and N-aminomethylation of indoles can be achieved using readily available potassium iodide as a catalyst and tert-butyl peroxybenzoate as a co-oxidant.
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
N-Methylaniline, ≥99%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
N-Methylaniline, 98%
Supelco
N-Methylaniline, analytical standard
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Gold etchant, standard