Merck
  • Copper(I)-catalyzed cascade dearomatization of 2-substituted tryptophols with iodonium [corrected] salts.
Copper(I)-catalyzed cascade dearomatization of 2-substituted tryptophols with iodonium [corrected] salts.
Organic letters (2012-08-16)
Chuan Liu, Wei Zhang, Li-Xin Dai, Shu-Li You
ABSTRACT
Copper(I)-catalyzed dearomative arylation and vinylation of 2-substituted tryptophols were realized with a subsequent cyclization reaction. The cascade dearomatization sequence provided versatile furoindoline derivatives with two quaternary carbon centers in good to excellent yields.
Sigma-Aldrich
3-(2-Hydroxyethyl)indole, 97%
Pricing and availability is not currently available.