Merck
  • Involvement of NMDA receptors in the antidepressant-like action of adenosine.
Involvement of NMDA receptors in the antidepressant-like action of adenosine.
Pharmacological reports : PR (2012-07-21)
Manuella P Kaster, Daniele G Machado, Adair R S Santos, Ana Lúcia S Rodrigues
ABSTRACT
In this work, the contribution of NMDA receptors to the antidepressant-like effect of adenosine in the forced swimming test (FST) was investigated. The pretreatment of mice with NMDA or D-serine was able to prevent the anti-immobility effect of either adenosine or MK-801 in the FST. In addition, the administration of a sub-effective dose of adenosine produced a synergistic effect with sub-effective doses of MK-801, ketamine and zinc chloride. Moreover, the immobility time of the mice treated with active doses of adenosine or N(6)-cyclohexyladenosine (CHA) plus MK-801 was not significantly different from that obtained with adenosine, CHA and MK-801 alone; by contrast, the combination between active doses of adenosine and CHA plus an active dose of the tricyclic antidepressant imipramine produced a greater effect in the FST than the administration of either drug alone. Together, the results suggest that the effect of adenosine in the FST is likely dependent on the inhibition of NMDA receptors mediated by the activation of adenosine A(1) receptors.
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride 0.1 M solution