Skip to Content
Merck
All Photos(3)

Key Documents

229997

Sigma-Aldrich

Zinc chloride

99.999% trace metals basis

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 Pa ( 305 °C)(lit.)
1 mmHg ( 428 °C)

Quality Level

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤15.0 ppm Trace Metal Analysis

bp

732 °C (lit.)

mp

293 °C (lit.)

solubility

H2O: very soluble(lit.)
acetone: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride is a strongly hygroscopic deliquescentsolid widely used as a Lewis acid catalyst in a variety of organictransformations.

Application

Zinc chloride is used as a catalyst:
  • in Knoevenagel condensation of carbonyl substrates with acidic methylene reagents
  • in solvent-free synthesis of dihydropyrimidinones.
It may be used as chemical activating agents, zinc chloride in particular is the most widely used chemical in the preparation of activated carbon.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A low cost adsorbent from agricultural waste corn cob by zinc chloride activation
Tsai WT, et al.
Bioresource Technology, 64(3), 211-217 (1998)
A highly efficient solvent-free synthesis of dihydropyrimidinones catalyzed by zinc chloride.
Sun Q, et al.
Synthesis, 07, 1047-1051 (2004)
Zinc chloride as a new catalyst for Knoevenagel condensation.
Shanthan RP and Venkataratnam RV.
Tetrahedron Letters, 32(41), 5821-5822 (1991)
Romaric Magerand et al.
Scientific reports, 11(1), 4528-4528 (2021-02-27)
Deinococcus bacteria are extremely resistant to radiation and other DNA damage- and oxidative stress-generating conditions. An efficient SOS-independent response mechanism inducing expression of several DNA repair genes is essential for this resistance, and is controlled by metalloprotease IrrE that cleaves
Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service