Merck
  • Utilization of a deuterated derivatization agent to synthesize internal standards for gas chromatography-tandem mass spectrometry quantification of silylated metabolites.
Utilization of a deuterated derivatization agent to synthesize internal standards for gas chromatography-tandem mass spectrometry quantification of silylated metabolites.
Journal of chromatography. A (2012-06-15)
Stina K Lien, Hans Fredrik Nyvold Kvitvang, Per Bruheim
ABSTRACT
GC-MS analysis of silylated metabolites is a sensitive method that covers important metabolite groups such as sugars, amino acids and non-amino organic acids, and it has become one of the most important analytical methods for exploring the metabolome. Absolute quantitative GC-MS analysis of silylated metabolites poses a challenge as different metabolites have different derivatization kinetics and as their silyl-derivates have varying stability. This report describes the development of a targeted GC-MS/MS method for quantification of metabolites. Internal standards for each individual metabolite were obtained by derivatization of a mixture of standards with deuterated N-methyl-N-trimethylsilyltrifluoroacetamide (d9-MSTFA), and spiking this solution into MSTFA derivatized samples prior to GC-MS/MS analysis. The derivatization and spiking protocol needed optimization to ensure that the behaviour of labelled compound responses in the spiked sample correctly reflected the behaviour of unlabelled compound responses. Using labelled and unlabelled MSTFA in this way enabled normalization of metabolite responses by the response of their deuterated counterpart (i.e. individual correction). Such individual correction of metabolite responses reproducibly resulted in significantly higher precision than traditional data correction strategies when tested on samples both with and without serum and urine matrices. The developed method is thus a valuable contribution to the field of absolute quantitative metabolomics.
Product Description
Supelco
MSTFA Reagent, ampule of 10 × 1.2 mL, analytical standard, Cerilliant®
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥98.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, for silylations, LiChropur